Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C7H5BClFO3/c9-6-2-7(10)5(8(12)13)1-4(6)3-11/h1-3,12-13H
Inchi Key
OOTMYKBNPFCNBY-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (4-Chloro-2-fluoro-5-formylphenyl)boronic acid
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
