Skip to Content
Merck

BDPH9BCCBBB5

Bide Pharmatech

3-(2-Fluorophenoxy)azetidine hydrochloride

Synonym(s):

3-(2-fluorophenoxy)azetidine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1N1Cl1F1
CAS Number:
Molecular Weight:
203.64
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid

Inchi Code

1S/C9H10FNO.ClH/c10-8-3-1-2-4-9(8)12-7-5-11-6-7;/h1-4,7,11H,5-6H2;1H

Inchi Key

TXHNDAUXWXARCI-UHFFFAOYSA-N



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library