Skip to Content
Merck

BDPH9BCD641A

Bide Pharmatech

4-(Methanesulfonyl)benzoyl chloride

Synonym(s):

4-(methylsulfonyl)benzoyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O3Cl1S1
CAS Number:
Molecular Weight:
218.66
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7ClO3S/c1-13(11,12)7-4-2-6(3-5-7)8(9)10/h2-5H,1H3

Inchi Key

IPEIDGXNXFPGBG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library