Skip to Content
Merck

BDPH9BCDB2FA

Bide Pharmatech

3,5-Dimethyl-1H-pyrrole-2-carboxylic acid

Synonym(s):

3,5-dimethyl-1H-pyrrole-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H9O2N1
CAS Number:
Molecular Weight:
139.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H9NO2/c1-4-3-5(2)8-6(4)7(9)10/h3,8H,1-2H3,(H,9,10)

Inchi Key

XQGVQRPSTODODM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library