Skip to Content
Merck

Skip To

BL3H160C1917

BLD Pharmatech

(2-Amino-6-bromopyridin-3-yl)methanol

Synonym(s):

(2-amino-6-bromopyridin-3-yl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Ships in 10 days - from Aldrich Partner.
€118.00
250 mg
Ships in 10 days - from Aldrich Partner.
€193.00
1 g
Ships in 10 days - from Aldrich Partner.
€540.00

About This Item

Linear Formula:
C6H7O1N2Br1
CAS Number:
Molecular Weight:
203.04
MDL number:

Vendor SKU: BD473346-100MG

€118.00


Ships in 10 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H7BrN2O/c7-5-2-1-4(3-10)6(8)9-5/h1-2,10H,3H2,(H2,8,9)

Inchi Key

QTTCZUHBAQUDIE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library