Skip to Content
Merck

Skip To

BPS60312

PDE1C Assay Kit

Synonym(s):

Assay Kits, PDE1C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121811
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

technique(s)

activity assay: suitable (Fluorescence Polarization)

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

The PDE1C Assay Kit is designed for identification of PDE1C inhibitors using fluorescence polarization. The assay is based on the binding of a fluorescent nucleotide monophosphate generated by PDE1C to the binding agent. The key to the PDE1C Assay Kit is the specific binding agent. Using this kit, only two simple steps on a microtiter plate are required for PDE1C reactions. First, the fluorescently labeled cAMP is incubated with a sample containing PDE1C for 1 hour. Second, a binding agent is added to the reaction mix to produce a change in fluorescent polarization. The FP signal is measured using a fluorescent microplate reader capable of measuring fluorescence polarization.

Application

Useful for studying enzyme kinetics and screening small molecular inhibitors for drug discovery and HTS applications.
Phosphodiesterases (PDEs) play an important role in the dynamic regulation of the second messengers cAMP (cyclic adenosine monophosphate) and cGMP (cyclic guanosine monophosphate) signaling, by hydrolyzing them. The PDE superfamily is composed of 11 families, with PDE4, 7 and 8 being cAMP-specific hydrolases, and thus regulating positive and negative responses to it. Among the 11 different families of PDEs, the phosphodiesterase 1 (PDE1) sub-family of enzymes hydrolyze both cAMP and cGMP in a mutually competitive manner. The catalytic activity of PDE1 is stimulated by their binding to Ca2+/calmodulin, resulting in the integration of Ca2+ and cyclic nucleotide-mediated signaling in various diseases. The PDE1 family includes three subtypes, PDE1A, PDE1B and PDE1C, which differ for their relative affinities for cAMP and cGMP. These isoforms are differentially expressed throughout the body, including the cardiovascular, central nervous system and other organs. PDE1 increases vascular contraction of mesenteric arteries upon angiotensin II infusion in rats and also regulates cGMP hydrolysis in cardiomyocyte and vascular smooth muscle cells, which in turn controls smooth muscle tone. Inhibition of PDE1 attenuates adverse cardiac remodeling and may serve as a novel therapeutic approach to limit nitrate tolerance.
Datasheet and Protocols may be found here.

Features and Benefits

The PDE1C inhibitor screening assay kit comes in a convenient 96-well format, including purified PDE1C enzyme, fluorescently labeled PDE1C substrate (cAMP), binding agent, and PDE assay buffer for 100 enzyme reactions.


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library