Skip to Content
Merck

ZRB2235

Anti-LYVE1 Antibody, clone 2F9 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cell surface retention sequence-binding protein 1 (CRSBP-1), Extracellular link domain-containing protein 1, Hyaluronic acid receptor., Lymphatic vessel endothelial hyaluronic acid receptor 1

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 35.21 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG

epitope sequence

C-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... LYVE1(10894)

General description

Lymphatic vessel endothelial hyaluronic acid receptor 1 (UniProt: Q9Y5Y7; LYVE-1, Cell surface retention sequence-binding protein 1, CRSBP-1, Extracellular link domain-containing protein 1, Hyaluronic acid receptor) is encoded by the LYVE1 (also known as CRSBP1, HAR, XLKD1, UNQ230/PRO263) gene (Gene ID: 10894) in human. LYVE-1 is a disulfide-linked, homodimeric, single-pass type I membrane protein that undergoes ligand-dependent internalization and recycling at the cell surface. It serves as a ligand-specific transporter trafficking between intracellular organelles (TGN) and the plasma membrane. It is synthesized with a signal peptide (aa 19), which is subsequently cleaved off to produce the mature protein that contains a large extracellular domain (aa 20-238), a transmembrane Helical domain (aa 239-259), and a cytoplasmic tail (aa 260-322). It is mainly expressed in endothelial cells lining lymphatic vessels. It is expressed on both the luminal and basolateral surfaces of lymphatic endothelium. It is reported to mediate the uptake of intermediate and high molecular weight hyaluronan (HA) and serves as a low affinity receptor for HA-induced lymphoangiogenesis. It also controls the entry and/or migration of leukocytes in lymphatic vessels. The capacity of LYVE-1 to bind HA is based on a receptor distribution and organization of the glycosaminoglycans. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lawrence, W et al. (2016). J Biol Chem. 291(15): 8014-8030).
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of extracellular domain of human LYVE1.

Application

Quality Control Testing

Evaluated by Immunohistochemistry by human liver tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected LYVE1 in human liver tissue sections.

Tested Applications

Flow Cytometry Analysis: 1 µg from a representative lot detected LYVE1 in one million human umbilical vein endothelial cells (HUVEC).

Affinity Binding Assay: A representative lot of this antibody bound LYVE1 peptide with at least one hundred thousand-fold (100,000X) higher affinity than with non-specific control peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LYVE1 in MCF7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 2F9 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects LYVE1. It targets an epitope within 21 amino acids from C-terminal region of extracellular domain.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service