Skip to Content
Merck

ZRB3092

Anti-Exportin-1/CRM1 Antibody, clone 1B2 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Chromosome region maintenance 1 protein homolog, Exp1

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 123.39 kDa
observed mol wt ~110 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... XPO1(7514)

General description

Exportin-1 (UniProt O14980; also known as Chromosome region maintenance 1 protein homolog, CRM1 homolog, Exp1) is encoded by the XPO1 (also known as CRM1) gene (Gene ID 7514) in human. It is a nuclear export protein that mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Most nuclear importins & exportins, including exportin-1, belong to the beta-karyopherin (KAP) family of proteins, Importins bind their cargoes with either nuclear localization signal (NLS) or M9 domain-containing hnRNP proteins in the cytoplasm, while exportins target their substrates via their nuclear export signal (NES) or M9 domain inside the nucleus. Structurally, XPO1 comprises multiple HEAT repeats that facilitate its interaction with cargo proteins bearing leucine-rich nuclear export signals (NES). Its expression is ubiquitous but is notably elevated in various malignancies, including hematological cancers and solid tumors. Exportin-1 is predominantly present in nucleus but shuttles between the nucleus/nucleolus and the cytoplasm. In the nucleus, Exportin-1 forms a complex with its cargo and the GTPase Ran in its active GTP-bound form, enabling efficient docking at the nuclear pore complex for transport. Exportin-1 plays a significant role in maintaining cellular homeostasis and regulating key processes such as the cell cycle and apoptosis. The upregulation and mutation of XPO1, such as the E571K variant, have been implicated in the pathogenesis of several cancers, making it a promising target for therapeutic intervention. Inhibitors of XPO1, such as selinexor, have shown efficacy in clinical trials, providing a novel strategy for cancer treatment by re-establishing the nuclear localization of tumor suppressor proteins and enhancing anti-tumor immunity. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ge, T., et al. (2025). Mol Cell. 85(6):1101-1116; Caillot, M., et al. (2023). Mol Oncol. 17(12):2546-2564).
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal half of human Exportin-1/CRM1.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Exportin-1/CRM1 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Exportin-1/CRM1 in lysate from MCF7 cells.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Exportin-1/CRM1 in HeLa cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Exportin-1/CRM1 in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1B2 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects Exportin-1/CRM1. It targets an epitope within 18 amino acids from the C-terminal half.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service