Merck
[Chemical modification of heparin].
Bioorganicheskaia khimiia (2006-10-18)
I Iu Ponedel'kina, V N Odinokova, E S Lukina, T V Tiumkina, L M Khalilov, U M Dzhemilev
ABSTRACT
Heparin was modified at carboxyl groups by reaction with several pharmacologically important amino-containing compounds in aqueous medium in the presence of 1-ethyl-3-[3-(dimethylamino)propyl]carbodiimide. In dependence on the nature of the amine and the ratio of reagents, conjugates containing 36-100% amide and 0-25% isoureidocarbonyl groups were synthesized. Isoureidoarylamide groups are present, along with amide moieties, in the products of heparin modification by hydroxyl-containing aromatic amines. The conjugate of heparin with p-aminobenzoic acid contained oligomeric arylamide.
Sigma-Aldrich
1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide methiodide
Pricing and availability is not currently available.