Merck
  • Kinetic enzymatic method for automated determination of HDL cholesterol in plasma.
Kinetic enzymatic method for automated determination of HDL cholesterol in plasma.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie (1987-09-01)
J S Moshides
ABSTRACT
A sensitive fixed-time kinetic enzymatic method for the measurement of high density lipoprotein cholesterol is described, as adapted for the Cobas Bio centrifugal analyser (Hoffmann LaRoche). The Boehringer Mannheim cholesterol esterase/cholesterol oxidase/peroxidase/3,4-dichlorophenol kinetic reagent was modified by the inclusion of 2,4,6-tribromo-3-hydroxybenzoic acid which reacts with hydrogen peroxide and 4-aminophenazone to produce a quinone-imine dye with a greater molar absorptivity than that produced with phenol. The method has been developed for the determination of HDL fractions isolated with polyethylene glycol 6000, for which a reagent of high sensitivity is required. The method is linear to at least 3.88 mmol/l of HDL cholesterol and the coefficients of variation for within-run and day-to-day precision were less than 3.0%. Correlation of the kinetic HDL cholesterol method with an equilibrium method was good (R = 0.9980). The assay is rapid, inexpensive and large numbers of specimens can be processed conveniently.
Sigma-Aldrich
2,4,6-Tribromo-3-hydroxybenzoic acid, 97%
Pricing and availability is not currently available.