Merck
SPKMBCHRVLV002
Spare part, Mobius® Chrom, Pinch Valve
new
NO PCV 3/8"
Select a Size
1 EACH
£1,700.00
£1,700.00
Estimated to ship on22 December 2025Details
About This Item
UNSPSC Code:
23151800
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Quality Level
product line
Mobius®
packaging
pack of 1 unit
compatibility
for use with Mobius®
General description
This pinch valve is designed for Mobius® Chrom systems.This spare part is classified as ′Critical,′ meaning that it is crucial, for avoiding system downtime, to keep at least one in stock for immediate availability.
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany