Skip to Content
Merck

AMBH2D6F0DEB

Ambeed, Inc.

1-(Methoxymethyl)-1H-imidazole

Synonym(s):

1-(methoxymethyl)-1H-imidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AMBH2D6F0DEB is not currently sold in your country.

About This Item

Linear Formula:
C5H8O1N2
CAS Number:
Molecular Weight:
112.13
MDL number:
Product AMBH2D6F0DEB is not currently sold in your country.

Inchi Code

1S/C5H8N2O/c1-8-5-7-3-2-6-4-7/h2-4H,5H2,1H3

Inchi Key

XJGYZYRJUUMCAP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless or White to Yellow Solid or Liquid

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library