Skip to Content
Merck

E5014

Benzonase ® Salt Tolerant Endonuclease

≥200 U/uL, for nucleic acid removal at high salt concentration

Synonym(s):

Endonuclease, nuclease

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352204

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

Escherichia coli (recombinant, expressed in E.coli)

Quality Level

form

buffered aqueous glycerol solution

mol wt

calculated mol wt 27 kDa

concentration

≥200 units/μL

storage temp.

-10 to -25°C

General description

The best-in-class solution for nucleic acid removal at high salt concentration. The Benzonase® Salt Tolerant endonuclease is an enzyme, different in amino acids sequence from other Benzonase® endonuclease references. It was designed with state-of-the-art protein engineering capabilities, to ensure the highest activity at high salt concentrations. The protein is a monomer with a molecular weight of about 27 kDa . The protein has an isoelectric point (pI) at pH 9.68. It is functional between pH 6 and 10, and from 0 °C to above 37 °C. Mg2+ (1–2 mM) is required for enzyme activity.

Application

Benzonase® Salt Tolerant endonuclease degrades both DNA and RNA, whether single stranded, double-stranded, linear, circular or supercoiled. No base preference is observed. As with all endonucleases, Benzonase® Salt Tolerant endonuclease hydrolyzes internal phosphodiester bonds present between the nucleotides. Upon complete digestion, all free nucleic acids present in solution are reduced to 5’-monophosphate terminated oligonucleotides, which are three to five bases in length.

Features and Benefits

  • Active at higher salt concentration, up to1M NaCl
  • Non-animal origin, recombinant from E.Coli, High Purity
  • No post-translational modifications
  • High batch to batch reproducibility
  • Free of detectable protease activity
  • Does not contain any antimicrobial preservatives or protein stabilizers except glycerol
  • Compatible with Deviron® detergents for cell lysis

Preparation Note

-10°C to -25°C

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
DEVIRON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service