Select a Size
About This Item
Skip To
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
hGB6, monoclonal
species reactivity
human
packaging
antibody small pack of 100 μg
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
isotype
IgMκ
epitope sequence
Unknown
shipped in
dry ice
target post-translational modification
unmodified
1 of 4
This Item | MABS2215 | MABS2190 | MABS2231 |
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified antibody | antibody form purified antibody | antibody form purified antibody | antibody form purified antibody |
| biological source mouse | biological source mouse | biological source mouse | biological source rat |
| species reactivity human | species reactivity human | species reactivity human | species reactivity mouse |
| clone hGB6, monoclonal | clone hGB1, monoclonal | clone hGB10, monoclonal | clone F5C12, monoclonal |
| shipped in dry ice | shipped in ambient | shipped in ambient | shipped in ambient |
General description
Immunogen
Application
Evaluated by Immunohistochemistry (Paraffin) in human gallbladder tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected all mucosal epithelium, a subset of lamina propria, peribiliary glands and surrounding connective tissue in human gallbladder tissue sections.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A representative lot detected Extrahepatic biliary cells in Immunohistochemistry applications (Galivo, F.H., et al. (2015). Stem Cell Res. 15(1):172-81).
Immunofluorescence Analysis: A representative lot detected Extrahepatic biliary cells in Immunofluorescence applications (Galivo, F.H., et al. (2015). Stem Cell Res. 15(1):172-81).
Flow Cytometry Analysis: A representative lot detected Extrahepatic biliary cells in Flow Cytometry applications (Galivo, F.H., et al. (2015). Stem Cell Res. 15(1):172-81).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


