Merck
AXYPCR32V
Corning® Axygen® PCR Microplate
size 32 wells, conical bottom violet polypropylene wells, pkg of 5x10plates/cs
Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NB.22
UNSPSC Code:
41121800
Material:
conical bottom violet polypropylene wells
Size:
32 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Product Name
Corning® Axygen® PCR Microplate, size 32 wells, conical bottom violet polypropylene wells, pkg of 5x10plates/cs
material
conical bottom violet polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-32-V
Corning
packaging
pkg of 5x10plates/cs
maximum volume
200 μL
size
32 wells
working volume
200 μL
suitability
suitable for (thermal cycling)
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)