Skip to Content
Merck

442517

Chlorobenzene-d5

analytical standard

Synonym(s):

Pentadeuterochlorobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C6D5Cl
CAS Number:
Molecular Weight:
117.59
EC Number:
221-482-0
UNSPSC Code:
12181503
PubChem Substance ID:
Beilstein/REAXYS Number:
1866335
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Chlorobenzene-d5, analytical standard

InChI key

MVPPADPHJFYWMZ-RALIUCGRSA-N

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

SMILES string

[2H]c1c([2H])c([2H])c(Cl)c([2H])c1[2H]

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.522 (lit.)

bp

130-130.5 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+5

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

75.2 °F - closed cup

flash_point_c

24.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service