Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
Empirical Formula (Hill Notation):
C42H45ClN2O2
Molecular Weight:
645.27
UNSPSC Code:
12352211
biological source
synthetic
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 10 mg (880184P-10MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
fluorescence
λex 785 nm; λem 820 nm
lipid type
fluorescent lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Cl-].CN1\C(=C\C=C\C=C\C=C\C2=[N+](CCCCCC(O)=O)c3ccc4ccccc4c3C2(C)C)C(C)(C)c2c1ccc1ccccc21
Related Categories
General description
Cyanine 7.5, free acid is a near-infrared fluorescent probe that can be used to label biomolecules such as peptides.
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service