Skip to Content
Merck

880245P

Avanti

DSG-PEG2000-DBCO

new

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-rac-glycero-3-[dibenzocyclooctyl(polyethylene glycol)-2000]

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C148H270N2O51
Molecular Weight:
2893.71
UNSPSC Code:
12352211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (880245P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)CCC(=O)N1Cc2ccccc2C#Cc2ccccc12)OC(=O)CCCCCCCCCCCCCCCCC

General description

DSG-PEG2000-DBCO is a lipid that consists of a DSG-PEG2000 unit conjugated to a DBCO unit. This lipid incorporates itself into micelles and liposomes.

Features and Benefits

The ability for this lipid to be clickable and can be conjugated to a variety of molecules using click chemistry.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service