Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
Empirical Formula (Hill Notation):
C36H41N3O6 · HCl
CAS Number:
Molecular Weight:
648.19
MDL number:
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
O=C(OC)C1=C(C)NC(C)=C(C(OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)=O)C1C4=CC([N+]([O-])=O)=CC=C4.Cl
InChI
1S/C36H41N3O6.ClH/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27;/h7-19,22,30,33,37H,20-21,23H2,1-6H3;1H
InChI key
WMFYOYKPJLRMJI-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Biochem/physiol Actions
Lercanidipine hydrochloride is a L-type (Cav1.2b) vascular channel antagonist; L-type (Cav1.2a) cardiac channel agonist voltage-dependent and highly lipophylic compound, which exhibits a slower onset and longer duration of action than other calcium channel antagonists; an antihypertensive agent in patients with mild-to-moderate hypertension; more vasoselective than lacidipine and amlodipine.
Lercanidipine hydrochloride is a L-type (Cav1.2b) vascular channel antagonist; L-type (Cav1.2a) cardiac channel agonist.
Packaging
Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service