Skip to Content
Products
Cart
0
ID
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Denhardt’s Solution
Synonym(s):
Denhardt′s Solution For Blotting Technique
Products
Product Category
buffers
Brand
Sigma-Aldrich
Physical Form
Quality Segment
Special Grade
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D9905
Denhardt’s Solution,
for Northern and Southern blotting, lyophilized powder
Expand
View Pricing
An unknown error has occured.