Skip to Content
Products
Cart
0
ID
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Electrolytic copper (trace elements)
Synonym(s):
Copper
Linear Formula:
Cu
CAS Number:
7440-50-8
Molecular Weight:
63.55
Products
Product Category
reference materials
Manufacturer
Special Grade
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
ERMEB074A
Electrolytic copper (trace elements),
ERM
®
, certified reference material
Expand
View Pricing
An unknown error has occured.