Skip to Content
Products
Cart
0
ID
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
pH Test
Synonym(s):
pH Visual Test Kit
Products
Product Category
Colorimetric & Titrimetric Test Kits
test strips
Brand
Supelco
Detection Method
Quality Segment
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.08027
pH Test,
pH range 4.5-9.0, graduations and accuracy: 0.5 pH unit
Expand
View Pricing
An unknown error has occured.