Merck
  • Voltage-dependent activation of Rac1 by Nav 1.5 channels promotes cell migration.
Voltage-dependent activation of Rac1 by Nav 1.5 channels promotes cell migration.
Journal of cellular physiology (2019-10-16)
Ming Yang, Andrew D James, Rakesh Suman, Richard Kasprowicz, Michaela Nelson, Peter J O'Toole, William J Brackenbury
ABSTRACT
Ion channels can regulate the plasma membrane potential (Vm ) and cell migration as a result of altered ion flux. However, the mechanism by which Vm regulates motility remains unclear. Here, we show that the Nav 1.5 sodium channel carries persistent inward Na+ current which depolarizes the resting Vm at the timescale of minutes. This Nav 1.5-dependent Vm depolarization increases Rac1 colocalization with phosphatidylserine, to which it is anchored at the leading edge of migrating cells, promoting Rac1 activation. A genetically encoded FRET biosensor of Rac1 activation shows that depolarization-induced Rac1 activation results in acquisition of a motile phenotype. By identifying Nav 1.5-mediated Vm depolarization as a regulator of Rac1 activation, we link ionic and electrical signaling at the plasma membrane to small GTPase-dependent cytoskeletal reorganization and cellular migration. We uncover a novel and unexpected mechanism for Rac1 activation, which fine tunes cell migration in response to ionic and/or electric field changes in the local microenvironment.
Sigma-Aldrich
Veratridine, ≥90% (HPLC), powder
Pricing and availability is not currently available.