Skip to Content
Merck
3-Butylidenephthalide phyproof® Reference Substance

3-Butylidenephthalide

3-Butylidenephthalide phyproof® Reference Substance
Synonym(s):
Butylidenephtalide, Ligusticum lactone, NSC 325307
Empirical Formula (Hill Notation):
C12H12O2
CAS Number:
Molecular Weight:
188.22

Products

Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing