Merck
  • Downregulation of CUEDC2 prevents doxorubicin‑induced cardiotoxicity in H9c2 cells.
Downregulation of CUEDC2 prevents doxorubicin‑induced cardiotoxicity in H9c2 cells.
Molecular medicine reports (2018-05-31)
Xianpu Zhang, Jiaojiao Li, Yongbo Cheng, Jianguang Yi, Xin Liu, Wei Cheng
ABSTRACT
Treatment with doxorubicin (DOX), which is an effective anticancer agent, is limited by cardiotoxicity. CUE domain‑containing 2 (CUEDC2) serves a role in numerous cellular processes. The present study aimed to elucidate the potential function of CUEDC2 in DOX‑induced cardiotoxicity. Cell Counting kit‑8 assay demonstrated that DOX induced cytotoxicity of H9c2 cells in a dose‑dependent manner. Flow cytometry demonstrated that downregulation of CUEDC2 reduced the levels of DOX‑induced reactive oxygen species. Furthermore, compared with in the DOX‑treated group, the activity of superoxide dismutase was increased in the DOX + small interfering RNA (si)CUEDC2 group; whereas, the malondialdehyde content was reduced in the DOX + siCUEDC2 group. In addition, flow cytometric analysis indicated that mitochondrial membrane potential was maintained following the depletion of CUEDC2. Furthermore, CUEDC2 downregulation significantly inhibited DOX‑induced apoptosis. The expression levels of proapoptotic genes, including B‑cell lymphoma 2 (Bcl‑2)‑associated X protein, cleaved caspase‑3 and cytochrome c were inhibited by the depletion of CUEDC2. Conversely, the expression levels of the anti‑apoptotic gene Bcl‑2 were elevated in the CUEDC2 knockdown group. Downregulation of CUEDC2 also increased phosphorylation of protein kinase B and forkhead box O3a, and decreased the expression of Bcl‑2‑like protein 11 according to western blot analysis. Taken together, the present study demonstrated that CUEDC2 downregulation prevented DOX‑induced cardiotoxicity in H9c2 cells. Therefore, CUEDC2 may be a promising target for the prevention of DOX‑induced cardiotoxicity.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CUEDC2