Merck
  • Recognition of adenosine monophosphate and H2PO4- using zinc ensemble of new hexaphenylbenzene derivative: potential bioprobe and multichannel keypad system.
Recognition of adenosine monophosphate and H2PO4- using zinc ensemble of new hexaphenylbenzene derivative: potential bioprobe and multichannel keypad system.
Organic letters (2012-01-28)
Vandana Bhalla, Varun Vij, Manoj Kumar, Parduman Raj Sharma, Tandeep Kaur
ABSTRACT
Zinc ensemble of hexaphenylbenzene derivative 3 exhibits sensitive response toward adenosine monophosphate (AMP) and H(2)PO(4)(-) ions. Further, the application of derivative 3 as a multichannel molecular keypad could be realized in the presence of inputs of Zn(2+) ions, H(2)PO(4)(-) ions, and AMP.
Sigma-Aldrich
4-Aminophenylboronic acid pinacol ester, 97%
Pricing and availability is not currently available.