Merck
  • Contrasting roles for domain 4 of VCAM-1 in the regulation of cell adhesion and soluble VCAM-1 binding to integrin alpha4beta1.
Contrasting roles for domain 4 of VCAM-1 in the regulation of cell adhesion and soluble VCAM-1 binding to integrin alpha4beta1.
Journal of immunology (Baltimore, Md. : 1950) (2006-04-06)
Darren G Woodside, Ronda M Kram, Jason S Mitchell, Tracie Belsom, Matthew J Billard, Bradley W McIntyre, Peter Vanderslice
ABSTRACT
Cell adhesion mediated by the interaction between integrin alpha4beta1 and VCAM-1 is important in normal physiologic processes and in inflammatory and autoimmune disease. Numerous studies have mapped the alpha4beta1 binding sites in VCAM-1 that mediate cell adhesion; however, little is known about the regions in VCAM-1 important for regulating soluble binding. In the present study, we demonstrate that 6D VCAM-1 (an alternatively spliced isoform of VCAM-1 lacking Ig-like domain 4) binds alpha4beta1 with a higher relative affinity than does the full-length form of VCAM-1 containing 7 Ig-like extracellular domains (7D VCAM-1). In indirect binding assays, the EC50 of soluble 6D VCAM-1 binding to alpha4beta1 on Jurkat cells (in 1 mM MnCl2) was 2 x 10(-9) M, compared with 7D VCAM-1 at 11 x 10(-9) M. When used in solution to inhibit alpha4beta1 mediated cell adhesion, the IC50 of 6D VCAM-1 was 13 x 10(-9) M, compared with 7D VCAM-1 measured at 150 x 10(-9) M. Removal of Ig-like domains 4, 5, or 6, or simply substituting Asp328 in domain 4 of 7D VCAM-1 with alanine, caused increased binding of soluble 7D VCAM-1 to alpha4beta1. In contrast, cells adhered more avidly to 7D VCAM-1 under shear force, as it induced cell spreading at lower concentrations than did 6D VCAM-1. Finally, soluble 6D VCAM-1 acts as an agonist through alpha4beta1 by augmenting cell migration and inducing cell aggregation. These results indicate that the domain 4 of VCAM-1 plays a contrasting role when VCAM-1 is presented in solution or as a cell surface-expressed adhesive substrate.
Sigma-Aldrich
Monoclonal Anti-Vascular Cell Adhesion Molecule 1 antibody produced in mouse, ~1 mg/mL, clone 1.4C3, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.