Merck
A short synthesis of (+)-cyclophellitol.
The Journal of organic chemistry (2005-11-19)
Flemming Gundorph Hansen, Eva Bundgaard, Robert Madsen
ABSTRACT
[reaction: see text] A new synthesis of (+)-cyclophellitol, a potent beta-glucosidase inhibitor, has been completed in nine steps from D-xylose. The key transformations involve a zinc-mediated fragmentation of benzyl-protected methyl 5-deoxy-5-iodo-xylofuranoside followed by a highly diastereoselective indium-mediated coupling with ethyl 4-bromocrotonate. Subsequent ring-closing olefin metathesis, ester reduction, olefin epoxidation, and deprotection then afford the natural product. This constitutes the shortest synthesis of (+)-cyclophellitol reported to date.