Skip to Content

AABH97D2A00A

AA BLOCKS, INC.

2-(3-fluoro-4-methylphenoxy)ethan-1-amine

Synonym(s):

2-(3-fluoro-4-methylphenoxy)ethan-1-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H12O1N1F1
CAS Number:
Molecular Weight:
169.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H12FNO/c1-7-2-3-8(6-9(7)10)12-5-4-11/h2-3,6H,4-5,11H2,1H3

Inchi Key

VYAMQORDSBYFKG-UHFFFAOYSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library