Skip to Content
Merck

Skip To

GF20249131

Indium

wire reel, 2m, diameter 0.15mm, as drawn, 99.999%

Synonym(s):

Indium, IN005105

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
UNSPSC Code:
12352300
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.01 mmHg ( 25 °C)

assay

99.999%

form

wire

manufacturer/tradename

Goodfellow 202-491-31

resistivity

8.37 μΩ-cm

L × diam.

2 m × 0.15 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Indium




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters