Skip to Content
Merck

Skip To

GF43448564

Brass alloy, Cu63Zn37

foil, 50 x 50mm, 0.075mm thickness, half hard, light tight (lt)

Synonym(s):

CU020250

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.23
UNSPSC Code:
12352300


description

light tight (lt)

form

foil (half hard)

manufacturer/tradename

Goodfellow 434-485-64

size × thickness

50 mm × 50 mm × 0.075 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?

Explore all of our products under Brass alloy, Cu63Zn37




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters