Skip to Content
Merck

Skip To

GF44475813

Niobium

foil, thickness 0.025 mm, 25 mm diameter, purity 99.9%

Synonym(s):

NB000301, Columbium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141730
MDL number:
eCl@ss:
38180501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 444-758-13

resistivity

13-16 μΩ-cm, 20°C

diam.

25 mm

thickness

0.025 mm

diameter

25 mm

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF40172600GF23685790GF54892930
description

foil, thickness 0.025 mm, 25 mm diameter, purity 99.9%

description

rod, 100mm, diameter 42mm, annealed, 99.9%

description

foil, thickness 0.006 mm, 25 mm diameter, purity 99.9%

description

wire reel, 50m, diameter 0.5mm, annealed, 99.9%

form

foil

form

rod

form

foil

form

wire

assay

99.9%

assay

99.9%

assay

99.9%

assay

99.9%

manufacturer/tradename

Goodfellow 444-758-13

manufacturer/tradename

Goodfellow 401-726-00

manufacturer/tradename

Goodfellow 236-857-90

manufacturer/tradename

Goodfellow 548-929-30

bp

4742 °C (lit.)

bp

4742 °C (lit.)

bp

4742 °C (lit.)

bp

4742 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

mp

2468 °C (lit.)

mp

2468 °C (lit.)

mp

2468 °C (lit.)

mp

2468 °C (lit.)


Still not finding the right product?

Explore all of our products under Niobium


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters