Inchi Code
1S/C8H7BrN2O/c1-12-7-3-2-6(9)5-4-10-11-8(5)7/h2-4H,1H3,(H,10,11)
Inchi Key
SIKUFDVMQXXDET-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown Solid
Still not finding the right product?
Explore all of our products under 4-Bromo-7-methoxy-1H-indazole
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
