Skip to Content
Merck

BDPH9BCC9BC6

Bide Pharmatech

N2,N2-Dimethylpyridine-2,5-diamine

Synonym(s):

N~2~,N~2~-dimethyl-2,5-pyridinediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H11N3
CAS Number:
Molecular Weight:
137.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H11N3/c1-10(2)7-4-3-6(8)5-9-7/h3-5H,8H2,1-2H3

Inchi Key

OBOSXEWFRARQPU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid



Still not finding the right product?

Explore all of our products under N2,N2-Dimethylpyridine-2,5-diamine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library