Inchi Code
1S/C11H22N2O4S/c1-4-11(14)12-7-5-8-13(2,3)9-6-10-18(15,16)17/h4H,1,5-10H2,2-3H3,(H-,12,14,15,16,17)
Inchi Key
KVKJQOXYGGPBIW-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 3-((3-Acrylamidopropyl)dimethylammonio)propane-1-sulfonate
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
