Skip to Content

BDPH9BCCD92D

Bide Pharmatech

3-(tert-Butyldimethylsilyl)prop-2-yn-1-ol

Synonym(s):

3-[tert-butyl(dimethyl)silyl]-2-propyn-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H18O1Si1
CAS Number:
Molecular Weight:
170.33
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H18OSi/c1-9(2,3)11(4,5)8-6-7-10/h10H,7H2,1-5H3

Inchi Key

BSFBEXRZAZEBTM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library