Skip to Content
Merck

Skip To

BDPH9BCD1CE4

Bide Pharmatech

1,3-Dithiane-2-carboxylic acid

Synonym(s):

1,3-dithiane-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C5H8O2S2
CAS Number:
Molecular Weight:
164.25
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H8O2S2/c6-4(7)5-8-2-1-3-9-5/h5H,1-3H2,(H,6,7)

Inchi Key

BZBYEAWKJNCPLB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?

Explore all of our products under 1,3-Dithiane-2-carboxylic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library