Skip to Content

BDPH9BCD5E75

Bide Pharmatech

4-Chloro-5-methoxy-2-methylpyrimidine

Synonym(s):

4-chloro-2-methyl-5-pyrimidinyl methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7O1N2Cl1
CAS Number:
Molecular Weight:
158.59
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H7ClN2O/c1-4-8-3-5(10-2)6(7)9-4/h3H,1-2H3

Inchi Key

SEPGLLQTLDBFAY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library