Skip to Content

BDPH9BCD91C0

Bide Pharmatech

4-Amino-1-ethyl-N-methyl-1H-pyrazole-3-carboxamide

Synonym(s):

4-amino-1-ethyl-N-methyl-1H-pyrazole-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H12O1N4
CAS Number:
Molecular Weight:
168.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H12N4O/c1-3-11-4-5(8)6(10-11)7(12)9-2/h4H,3,8H2,1-2H3,(H,9,12)

Inchi Key

JEQXMZSIIMYFTN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library