Skip to Content
Merck

BDPH9BCDA854

Bide Pharmatech

Dimethyl 4,5-dihydroxyphthalate

Synonym(s):

dimethyl 4,5-dihydroxyphthalate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O6
CAS Number:
Molecular Weight:
226.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10O6/c1-15-9(13)5-3-7(11)8(12)4-6(5)10(14)16-2/h3-4,11-12H,1-2H3

Inchi Key

FXPCZDQRACWUBW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under Dimethyl 4,5-dihydroxyphthalate


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library