Skip to Content
Merck

BDPH9BCDBCCD

Bide Pharmatech

4-(4-Chlorobenzyl)piperidine hydrochloride

Synonym(s):

4-(4-chlorobenzyl)piperidine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H17N1Cl2
CAS Number:
Molecular Weight:
246.18
MDL number:
Product BDPH9BCDBCCD is not currently sold in your country.


Inchi Code

1S/C12H16ClN.ClH/c13-12-3-1-10(2-4-12)9-11-5-7-14-8-6-11;/h1-4,11,14H,5-9H2;1H

Inchi Key

CRYGKHFRQLLVHQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library