Skip to Content

BDPH9BCDCCC9

Bide Pharmatech

4-(2-Hydroxypropan-2-yl)benzaldehyde

Synonym(s):

4-(2-hydroxypropan-2-yl)benzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O2
CAS Number:
Molecular Weight:
164.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12O2/c1-10(2,12)9-5-3-8(7-11)4-6-9/h3-7,12H,1-2H3

Inchi Key

RCUMJJBGXUQSOF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library