Skip to Content

BDPH9BCE13EA

Bide Pharmatech

2-Bromo-1-(2-fluoro-4-methoxyphenyl)ethanone

Synonym(s):

2-bromo-1-(2-fluoro-4-methoxyphenyl)ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O2Br1F1
CAS Number:
Molecular Weight:
247.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H8BrFO2/c1-13-6-2-3-7(8(11)4-6)9(12)5-10/h2-4H,5H2,1H3

Inchi Key

OKLFPKDKQYVRNJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Yellow to Brown Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library