Skip to Content
Merck

BDPH9C021A7C

Bide Pharmatech

1-Methyl-10H-phenoselenazine

Synonym(s):

1-methyl-10H-phenoselenazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C021A7C is not currently sold in your country.


Inchi Code

1S/C13H11NSe/c1-9-5-4-8-12-13(9)14-10-6-2-3-7-11(10)15-12/h2-8,14H,1H3

Inchi Key

YALDOISQBIOCMB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1-Methyl-10H-phenoselenazine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library