Skip to Content
Merck

BDPH9C021CA3

Bide Pharmatech

MaP4343

Synonym(s):

(3beta)-3-methoxypregn-5-en-20-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H34O2
CAS Number:
Molecular Weight:
330.51
MDL number:
Product BDPH9C021CA3 is not currently sold in your country.


Inchi Code

1S/C22H34O2/c1-14(23)18-7-8-19-17-6-5-15-13-16(24-4)9-11-21(15,2)20(17)10-12-22(18,19)3/h5,16-20H,6-13H2,1-4H3/t16-,17-,18+,19-,20-,21-,22+/m0/s1

Inchi Key

ZVGQOQHAMHMMNE-BIBIXIOVSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under MaP4343



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library