Skip to Content
Merck

BDPH9C021F4D

Bide Pharmatech

1-Boc-D-prolyl-glycine

Synonym(s):

({[(2R)-1-(tert-butoxycarbonyl)-2-pyrrolidinyl]carbonyl}amino)acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H20O5N2
Molecular Weight:
272.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H20N2O5/c1-12(2,3)19-11(18)14-6-4-5-8(14)10(17)13-7-9(15)16/h8H,4-7H2,1-3H3,(H,13,17)(H,15,16)/t8-/m1/s1

Inchi Key

PLJXCLBLFPFXEN-MRVPVSSYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library