Skip to Content
Merck

Skip To

ABC151

Anti-Annexin A1, Antibody

from rabbit

Synonym(s):

Annexin A1, Annexin I, Annexin-1, Calpactin II, Calpactin-2, Chromobindin-9, Lipocortin I, Phospholipase A2 inhibitory protein, p35

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANXA1(301)

General description

Annexin A1 (ANXA1) is a calcium and phospholipid binding protein almost entirely restricted to microglia in the CNS. During the peripheral inflammatory response, Annexin A1 is instrumental in the phagocytosis of apoptotic leukocytes. Annexin A1 restores the ability of microglia to differentiate between apoptotic and non-apoptotic cells during inflammatory activation. Current research is geared towards using Annexin A1 as a therapeutic target in neuroinflammation. Annexin A1 has also been shown to be influential in cancer development and proliferation.
~38 kDa observed

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human Annexin A1.

Application

Anti-Annexin A1 is an antibody targeting the Annexin A1 protein, validated for use in WB & IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blot Analysis: 2.0 µg/mL from a representative lot detected Annexin A1 in 10 µg of C2C12, HUVEC, and NIH/3T3 cell lysate.
Immunohistochemistry Analysis: 1:500 dilution from a representative lot detected Annexin A1 in human lung tissue.

Biochem/physiol Actions

This antibody recognizes the C-terminus of Annexin A1

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
L6 cell lysate
Evaluated by Western Blot in L6 cell lysate.

Western Blot Analysis: 2.0 µg/mL of this antibody detected Annexin A1 in 10 µg of L6 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters