Skip to Content
Merck

Skip To

AV13018

Anti-CHRNA7 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cholinergic receptor, nicotinic, α 7

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

56 kDa

species reactivity

human, mouse, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CHRNA7(1139)

Immunogen

The immunogen for anti-CHRNA7 antibody: synthetic peptide derected towards the N terminal of human CHRNA7

Application

Anti-CHRNA7 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

CHRNA7 is a transmembrane, oligomeric, ligand-gated nicotinic receptor which upon activation by cholinergic binding induces ion channel opening for the movement of positive ions. Nicotinic acetylcholine receptors mediate presynaptic, postsynaptic and extrasynaptic signaling. CHRNA7 has a role in the release of dopamine from striatum and prefrontal cortex in rats indicating a role in the progression of Parkinson′s disease and schizophrenia.

Physical form

Lyophilized from PBS buffer with 2% sucrose

Other Notes

Synthetic peptide located within the following region: QGEFQRKLYKELVKNYNPLERPVANDSQPLTVYFSLSLLQIMDVDEKNQV

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters