Skip to Content

OGS1371

PSF-CMV-PURO-NH2-FLUC - N-TERMINAL FLUC TAG MAMMALIAN PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
firefly luciferase
Peptide cleavage:
no cleavage
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

firefly luciferase conjugate

form

buffered aqueous solution

mol wt

size 7796 bp

bacteria selection

kanamycin

mammalian cells selection

puromycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

peptide tag location

N-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

firefly luciferase

shipped in

ambient

storage temp.

−20°C

General description

This plasmid is designed to express tagged proteins in mammalian cells either by transient transfection or by creating stable cell lines. It contains a puromycin resistance expression cassette using the human Ubiquitin promoter to drive expression and allow for the selection of cells containing the plasmid.

About the Peptide Tag:This plasmid contains an n-terminal Firefly luciferase reporter tag that can be fused to a gene of interest to allow protein detection.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library