Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.85
UNSPSC Code:
12352200
eCl@ss:
32160703
Promoter:
Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast
Promoter activity: constitutive
Promoter type: yeast
Origin of replication:
2Micron, pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
firefly luciferase
Peptide cleavage:
3C
conjugate
firefly luciferase conjugate
tag
6-His tagged
form
buffered aqueous solution
mol wt
size 8406 bp
technique(s)
molecular cloning: suitable
bacteria selection
kanamycin
origin of replication
2Micron, pUC (500 copies)
peptide cleavage
3C
peptide tag location
C-terminal
promoter
Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast
reporter gene
firefly luciferase
shipped in
ambient
storage temp.
−20°C
yeast selection
uracil
General description
This plasmid is designed to express tagged proteins in yeast cells (Saccharomyces cerevisiae). The plasmid contains an auxotrophic Uracil selection expression cassette (URA3) that allows for the positive selection of yeast that are deficient in the URA3 gene (YEL021W). This is typically achieved by growing the yeast in minimal media that is reconstituted with the essential amino acids and nucleotides but excluding Uracil.
About the Peptide Tag:This plasmid contains a c-terminal Firefly luciferase reporter tag that can be fused to a gene of interest to allow protein detection. This plasmid also contains a secondary Hexa-Histidine (6His) tag protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.
About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a 3C cleavage tag. The protein sequence of the cleavage tag is: LEVLFQGP. Human Rhinovirus (HRV) 3C Protease is a highly specific protease that cleaves between the Glu and Gly residues of its recognition site. It is often produced with the trademname ′PreScission protease′.
Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.
About the Peptide Tag:This plasmid contains a c-terminal Firefly luciferase reporter tag that can be fused to a gene of interest to allow protein detection. This plasmid also contains a secondary Hexa-Histidine (6His) tag protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.
About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a 3C cleavage tag. The protein sequence of the cleavage tag is: LEVLFQGP. Human Rhinovirus (HRV) 3C Protease is a highly specific protease that cleaves between the Glu and Gly residues of its recognition site. It is often produced with the trademname ′PreScission protease′.
Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.
Other Notes
Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics™. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.
Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.
Genebank Vector Sequence File
FASTA Vector Sequence File
Full Plasmid Map
Genebank Vector Sequence File
FASTA Vector Sequence File
Full Plasmid Map
Legal Information
Oxford Genetics is a trademark of Oxford Genetics Ltd
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.